Please login to your online account to display your discounted pricing

Mouse anti-Helicobacter pylori, Clone: SPM526, Novus Biologicals™

$307.00 - $518.93

Specifications

Antigen Helicobacter pylori
Clone SPM526
Immunogen Purified helicobacter pylori
Formulation 10mM PBS, pH 7.4. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP13595601

 
novus biologicals canada
NBP13595601ML
0.1mL Each for $307.00

NBP13595610

 
novus biologicals canada
NBP13595610ML
1mL Each for $518.93
Description & Specifications

Specifications

Antigen Helicobacter pylori
Clone SPM526
Immunogen Purified helicobacter pylori
Formulation 10mM PBS, pH 7.4. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Bacteria
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)