Please login to your online account to display your discounted pricing

Mouse anti-Helicobacter pylori, Clone: SPM526, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP13595670ML

 View all options for this product


For Research Use Only

Catalog No. NBP13595670


Description & Specifications


Antigen Helicobacter pylori
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone SPM526
Conjugate Unlabeled
Cross Reactivity Bacteria
Format Affinity Purified
Formulation 10mM PBS, pH 7.4. Immunogen affinity purified.
Host Species Mouse
Immunogen Purified helicobacter pylori
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 7mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity This product is specific for H. pylori infected tissue.