Please login to your online account to display your discounted pricing

Mouse anti-Helicobacter pylori, Clone: SPM526, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13595601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13595601

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Antigen Helicobacter pylori
Format Affinity Purified
Isotype IgG1, κ
Primary or Secondary Primary
Conjugate Unlabeled
Formulation 10mM PBS, pH 7.4. Immunogen affinity purified.
Host Species Mouse
Immunogen Purified helicobacter pylori
Purification Method Affinity Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Test Specificity This product is specific for H. pylori infected tissue.
Cross Reactivity Bacteria
Clone SPM526