Please login to your online account to display your discounted pricing

Mouse anti-Helicobacter pylori, Clone: SPM526, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13595610ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13595610

  • / Each
 

Description & Specifications

Specifications

Antigen Helicobacter pylori
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone SPM526
Conjugate Unlabeled
Format Affinity Purified
Formulation 10mM PBS, pH 7.4. Immunogen affinity purified.
Host Species Murine
Immunogen Purified helicobacter pylori
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity This product is specific for H. pylori infected tissue.
Target Species Bacteria