Please login to your online account to display your discounted pricing

Mouse anti-HVEM/TNFRSF14, Clone: HVEM-122, Novus Biologicals™

$274.00 - $274.00

Specifications

Antigen HVEM/TNFRSF14
Clone HVEM-122
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was HVEM.
Gene ID (Entrez) 8764
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14326803

 
novus biologicals canada
NBP1432680025MG
0.025mg Each for $274.00
Description & Specifications

Specifications

Antigen HVEM/TNFRSF14
Clone HVEM-122
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was HVEM.
Gene ID (Entrez) 8764
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Western Blot