Please login to your online account to display your discounted pricing

Mouse anti-HVEM/TNFRSF14, Clone: HVEM-122, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1432680025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14326803

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Applications Flow Cytometry
Applications Western Blotting
Format Affinity Purified
Isotype IgG1
Quantity 0.025mg
Conjugate Unlabeled
Concentration 0.5mg/mL
Regulatory Status RUO
Gene ID (Entrez) 8764
Primary or Secondary Primary
Host Species Mouse
Immunogen The immunogen for this antibody was HVEM.
Antigen HVEM/TNFRSF14
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C. Do not freeze.
Purification Method Affinity Purified
Formulation PBS [pH 7.2]
Clone HVEM-122