Please login to your online account to display your discounted pricing

Mouse anti-HVEM/TNFRSF14, Clone: HVEM-122, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14326801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14326801

  • / Each
 

Description & Specifications

Specifications

Antigen HVEM/TNFRSF14
Applications Flow Cytometry
Applications Western Blotting
Clone HVEM-122
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Mouse
Immunogen The immunogen for this antibody was HVEM.
Isotype IgG1
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 8764