Please login to your online account to display your discounted pricing

Mouse anti-HSPA8/HSC71/Hsc70, Clone: plant, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP197498005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19749805

  • / Each
 

Description & Specifications

Specifications

Antigen HSPA8/HSC71/Hsc70
Applications Immunoprecipitation
Applications Western Blot
Clone plant
Conjugate Unlabeled
Format Purified
Formulation Liquid. In PBS containing 50% glycerol. Protein G purified.
Host Species Murine
Immunogen Recombinant spinach Hsc70 (Hsp73).
Isotype IgG1
Purification Method Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3312
Target Species Plant
Target Species Porcine