Please login to your online account to display your discounted pricing

Mouse anti-HspA6, Clone: 165f, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP197466005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19746605

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Applications Western Blotting
Clone 165f
Quantity 0.05mg
Cross Reactivity Human
Host Species Mouse
Format Purified
Regulatory Status RUO
Formulation Liquid. In PBS, pH 7.2 containing 50% glycerol. Protein G purified.
Immunogen Synthetic peptide corresponding to the sequence near the C-terminus of human HspA6 Antibody (165f).
Antigen HspA6
Purification Method Purified
Isotype IgG1
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3310
Conjugate Unlabeled