Please login to your online account to display your discounted pricing

Mouse anti-HSP90, Clone: AC88, PE, Novus Biologicals™

$406.00 - $494.00

Specifications

Antigen HSP90
Clone AC88
Immunogen Achlya ambisexualis (water mold) Hsp90.
Gene ID (Entrez) 3320
Formulation Liquid. In PBS. Protein G purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP19744902

 
novus biologicals canada
NBP19744902MG
0.2mg Each for $494.00

NBP19744905

 
novus biologicals canada
NBP197449005MG
0.05mg Each for $406.00
Description & Specifications

Specifications

Antigen HSP90
Clone AC88
Immunogen Achlya ambisexualis (water mold) Hsp90.
Gene ID (Entrez) 3320
Formulation Liquid. In PBS. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1
Conjugate PE
Format Purified
Purification Method Purified
Applications Flow Cytometry