Please login to your online account to display your discounted pricing

Mouse anti-HSP90, Clone: AC88, DyLight 488, Novus Biologicals™

$406.00 - $536.00

Specifications

Antigen HSP90
Clone AC88
Immunogen Achlya ambisexualis (water mold) Hsp90.
Gene ID (Entrez) 3320
Formulation Liquid. In PBS containing 50% glycerol. Protein G purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP19748402

 
novus biologicals canada
NBP19748402MG
0.2mg Each for $536.00

NBP19748405

 
novus biologicals canada
NBP197484005MG
0.05mg Each for $406.00
Description & Specifications

Specifications

Antigen HSP90
Clone AC88
Immunogen Achlya ambisexualis (water mold) Hsp90.
Gene ID (Entrez) 3320
Formulation Liquid. In PBS containing 50% glycerol. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1
Conjugate DyLight 488
Format Purified
Purification Method Purified
Applications Flow Cytometry