Please login to your online account to display your discounted pricing

Mouse anti-HSP90 beta, Clone: K3701, Novus Biologicals™

$310.66 - $518.93

Specifications

Antigen HSP90 beta
Clone K3701
Immunogen Recombinant human HSP90beta.
Gene ID (Entrez) 3326
Formulation Liquid. In saline, pH 6.9. Ammonium sulfate precipitation.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP19746205

 
novus biologicals canada
NBP197462005MG
0.05mg Each for $310.66

NBP19746202

 
novus biologicals canada
NBP19746202MG
0.2mg Each for $518.93
Description & Specifications

Specifications

Antigen HSP90 beta
Clone K3701
Immunogen Recombinant human HSP90beta.
Gene ID (Entrez) 3326
Formulation Liquid. In saline, pH 6.9. Ammonium sulfate precipitation.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Guinea Pig
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Isotype IgM
Conjugate Unlabeled
Format Purified
Purification Method Precipitation
Applications Flow Cytometry
Applications Western Blotting