Please login to your online account to display your discounted pricing

Mouse anti-HSP90 beta, Clone: K3701, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP197462005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19746205

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation Liquid. In saline, pH 6.9. Ammonium sulfate precipitation.
Host Species Mouse
Applications Flow Cytometry
Applications Western Blotting
Immunogen Recombinant human HSP90beta.
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Clone K3701
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Guinea Pig
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Antigen HSP90 beta
Isotype IgM
Purification Method Precipitation
Quantity 0.05mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3326