Please login to your online account to display your discounted pricing

Mouse anti-HSP90 beta, Clone: K3701, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP19746202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19746202

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Western Blotting
Format Purified
Formulation Liquid. In saline, pH 6.9. Ammonium sulfate precipitation.
Isotype IgM
Regulatory Status RUO
Gene ID (Entrez) 3326
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Guinea Pig
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Antigen HSP90 beta
Host Species Mouse
Immunogen Recombinant human HSP90beta.
Purification Method Precipitation
Quantity 0.2mg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Clone K3701
Monoclonal or Polyclonal Monoclonal