Please login to your online account to display your discounted pricing

Mouse anti-Hsp70 / Hsc70, Clone: N27F3-4, Novus Biologicals™

$309.05 - $582.00

Specifications

Antigen Hsp70 / Hsc70
Clone N27F3-4
Immunogen Purified Hsc70 and Hsp70 isolated from human HeLa cells.
Gene ID (Entrez) 3303
Formulation Liquid. In PBS containing 50% glycerol. Protein G purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP19786502

 
novus biologicals canada
NBP19786502MG
0.2mg Each for $582.00

NBP19786505

 
novus biologicals canada
NBP197865005MG
0.05mg Each for $309.05
Description & Specifications

Specifications

Antigen Hsp70 / Hsc70
Clone N27F3-4
Immunogen Purified Hsc70 and Hsp70 isolated from human HeLa cells.
Gene ID (Entrez) 3303
Formulation Liquid. In PBS containing 50% glycerol. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Fish
Cross Reactivity Guinea Pig
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Plant
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Xenopus
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting