Please login to your online account to display your discounted pricing

Mouse anti-Hsp70 / Hsc70, Clone: N27F3-4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP197865005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19786505

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen Hsp70 / Hsc70
Quantity 0.05mg
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Formulation Liquid. In PBS containing 50% glycerol. Protein G purified.
Gene ID (Entrez) 3303
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Fish
Cross Reactivity Guinea Pig
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Plant
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Xenopus
Primary or Secondary Primary
Immunogen Purified Hsc70 and Hsp70 isolated from human HeLa cells.
Purification Method Purified
Conjugate Unlabeled
Format Purified
Isotype IgG1
Clone N27F3-4
Host Species Mouse