Please login to your online account to display your discounted pricing

Mouse anti-HSP65 (mycobacterial), Clone: 4H11, Novus Biologicals™

$300.00 - $367.12

Specifications

Antigen HSP65 (mycobacterial)
Clone 4H11
Immunogen Recombinant M. bovis BCG Hsp65.
Gene ID (Entrez) 886354
Formulation Liquid. In PBS, pH 7.2 containing 50% glycerol. Protein G purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP19787425

 
novus biologicals canada
NBP1978740025MG
0.025mg Each for $300.00

NBP19787401

 
novus biologicals canada
NBP19787401MG
0.1mg Each for $367.12
Description & Specifications

Specifications

Antigen HSP65 (mycobacterial)
Clone 4H11
Immunogen Recombinant M. bovis BCG Hsp65.
Gene ID (Entrez) 886354
Formulation Liquid. In PBS, pH 7.2 containing 50% glycerol. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications In vitro Assay
Applications Western Blot