missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-HSP65 (mycobacterial), Clone: 4H11, Novus Biologicals™

Manufacturer:  Novus Biologicals Canada NBP1978740025MG

 View more versions of this product

Catalog No. NBP19787425


 

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

HSP65 (mycobacterial)
4H11
Purified
Murine
IgG1
0.025mg
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Bacteria,Bovine
In vitro Assay,Western Blot
Unlabeled
Liquid. In PBS, pH 7.2 containing 50% glycerol. Protein G purified.
Recombinant M. bovis BCG Hsp65.
Purified
RUO
Primary
886354