Please login to your online account to display your discounted pricing

Mouse anti-HSP65 (mycobacterial), Clone: 4H11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP19787401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19787401

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bacteria
Cross Reactivity Bovine
Format Purified
Formulation Liquid. In PBS, pH 7.2 containing 50% glycerol. Protein G purified.
Host Species Mouse
Applications In vitro Assay
Applications Western Blotting
Isotype IgG1
Regulatory Status RUO
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Antigen HSP65 (mycobacterial)
Immunogen Recombinant M. bovis BCG Hsp65.
Quantity 0.1mg
Primary or Secondary Primary
Gene ID (Entrez) 886354
Clone 4H11
Purification Method Purified