Please login to your online account to display your discounted pricing

Mouse anti-HSP60, Clone: SPM253, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen HSP60
Clone SPM253
Concentration 0.2mg/mL
Immunogen Recombinant human HSP60 protein
Gene Accession No. P10809
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23286601

 
novus biologicals canada
NBP23286601ML
0.1mL Each for $139.00

NBP23286610

 
novus biologicals canada
NBP23286610ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen HSP60
Clone SPM253
Concentration 0.2mg/mL
Immunogen Recombinant human HSP60 protein
Gene Accession No. P10809
Gene ID (Entrez) 3329
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Drosophila
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Xenopus
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting