Please login to your online account to display your discounted pricing

Mouse anti-HSP60, Clone: SPM253, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23286610ML

 View all options for this product


For Research Use Only

Catalog No. NBP23286610


Description & Specifications


Antigen HSP60
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM253
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Drosophila
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Xenopus
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P10809
Host Species Mouse
Immunogen Recombinant human HSP60 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3329
Test Specificity Does not react with Bacteria, helminths or spinach.
Reconstitution Protein A purified