Please login to your online account to display your discounted pricing

Mouse anti-HSP60, Clone: SPM253, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23286601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23286601

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mL
Test Specificity Does not react with Bacteria, helminths or spinach.
Regulatory Status RUO
Immunogen Recombinant human HSP60 protein
Gene Accession No. P10809
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Drosophila
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Xenopus
Gene ID (Entrez) 3329
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Antigen HSP60
Conjugate Unlabeled
Isotype IgG1, κ
Concentration 0.2mg/mL
Purification Method Purified
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Primary or Secondary Primary
Formulation PBS with 0.05% BSA.
Clone SPM253
Host Species Mouse
Format Purified