Please login to your online account to display your discounted pricing

Mouse anti-HSP60, Clone: SPM253, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23473402MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23473402

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Regulatory Status RUO
Immunogen Recombinant human HSP60 protein
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Drosophila
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Xenopus
Antigen HSP60
Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P10809
Isotype IgG1, κ
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Gene ID (Entrez) 3329
Concentration 1mg/mL
Conjugate Unlabeled
Quantity 0.2mg
Clone SPM253
Test Specificity Recognizes a 60kDa protein, identified as the heat shock protein 60 (hsp60). Its epitope is localized between aa 383-447 of human hsp60. Does not react with helminths or spinach.
Purification Method Purified