Please login to your online account to display your discounted pricing

Mouse anti-HSP60, Clone: SPM253, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23473401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23473401

  • / Each
 

Description & Specifications

Specifications

Antigen HSP60
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone SPM253
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P10809
Host Species Murine
Immunogen Recombinant human HSP60 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3329
Test Specificity Recognizes a 60kDa protein, identified as the heat shock protein 60 (hsp60). Its epitope is localized between aa 383-447 of human hsp60. Does not react with helminths or spinach.
Reconstitution Protein A purified
Target Species Bovine
Target Species Canine
Target Species Chicken
Target Species Drosophila
Target Species Hamster
Target Species Human
Target Species Murine
Target Species Porcine
Target Species Primate
Target Species Rabbit
Target Species Rat
Target Species Sheep
Target Species Xenopus