missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-HSP60, Clone: LK2, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen HSP60
Clone LK2
Concentration 1mg/mL
Immunogen Recombinant human HSP60 protein
Gene Accession No. P10809
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23467101 Novus Biologicals Canada
NBP23467101MG
0.1mg Each for $384.00
NBP23467102 Novus Biologicals Canada
NBP23467102MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

HSP60
1mg/mL
P10809
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
LK2
Recombinant human HSP60 protein
3329
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot