Please login to your online account to display your discounted pricing

Mouse anti-HSP60, Clone: LK2, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23467101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23467101

  • / Each
 

Description & Specifications

Specifications

Immunogen Recombinant human HSP60 protein
Gene ID (Entrez) 3329
Isotype IgG1, κ
Test Specificity Reacts with: E. coli, Borellia, Streptococcus pyogenes, Yersinia entercolitica, Salmonella typhimurium, Treponema hyodysenteriae, Treponema innocense, Trichinella spiralis, Helminths, Yeast, Spinach
Cross Reactivity Bacteria
Cross Reactivity Chicken
Cross Reactivity Fungi
Cross Reactivity Guinea Pig
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Quantity 0.1mg
Antigen HSP60
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Gene Accession No. P10809
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Clone LK2
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Regulatory Status RUO
Purification Method Purified
Concentration 1mg/mL
Conjugate Unlabeled