Please login to your online account to display your discounted pricing

Mouse anti-HSP60, Clone: LK1, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen HSP60
Clone LK1
Concentration 1mg/mL
Immunogen Recombinant human HSP60 protein
Gene Accession No. P10809
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23467001

 
novus biologicals canada
NBP23467001MG
0.1mg Each for $384.00

NBP23467002

 
novus biologicals canada
NBP23467002MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen HSP60
Clone LK1
Concentration 1mg/mL
Immunogen Recombinant human HSP60 protein
Gene Accession No. P10809
Gene ID (Entrez) 3329
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Drosophila
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Xenopus
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting