Please login to your online account to display your discounted pricing

Mouse anti-HSP60, Clone: LK1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23467002MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23467002

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Quantity 0.2mg
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Drosophila
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Xenopus
Immunogen Recombinant human HSP60 protein
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Clone LK1
Purification Method Purified
Reconstitution Protein A purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Accession No. P10809
Test Specificity Does not react with Bacteria, helminths or spinach.
Host Species Mouse
Gene ID (Entrez) 3329
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Regulatory Status RUO
Isotype IgG1, κ
Concentration 1mg/mL
Primary or Secondary Primary
Antigen HSP60