Please login to your online account to display your discounted pricing

Mouse anti-HSP60, Clone: LK1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23467001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23467001

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Drosophila
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Xenopus
Antigen HSP60
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Isotype IgG1, κ
Test Specificity Does not react with Bacteria, helminths or spinach.
Purification Method Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Clone LK1
Conjugate Unlabeled
Quantity 0.1mg
Format Purified
Immunogen Recombinant human HSP60 protein
Gene ID (Entrez) 3329
Reconstitution Protein A purified
Host Species Mouse
Concentration 1mg/mL
Gene Accession No. P10809
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.