Please login to your online account to display your discounted pricing

Mouse anti-HSP60, Clone: GROEL730, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23462502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23462502

  • / Each
 

Description & Specifications

Specifications

Antigen HSP60
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone GROEL730
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P10809
Host Species Murine
Immunogen Recombinant human HSP60 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3329
Test Specificity Does not react to Mouse or Rat.
Reconstitution Protein A purified
Target Species Human