Please login to your online account to display your discounted pricing

Mouse anti-HSP30, Clone: RT30.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP19787102MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19787102

  • / Each
 

Description & Specifications

Specifications

Applications Western Blotting
Purification Method Purified
Cross Reactivity Fish
Immunogen Native rainbow trout HSP30.
Format Purified
Antigen HSP30
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Clone RT30.1
Isotype IgG1
Host Species Mouse
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Quantity 0.2mg
Formulation Liquid. In PBS containing 50% glycerol. Protein G purified.
Regulatory Status RUO
Primary or Secondary Primary