Please login to your online account to display your discounted pricing

Mouse anti-HSP27, Clone: SPM252, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23286510ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23286510

  • / Each
 

Description & Specifications

Specifications

Antigen HSP27
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM252
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Chimpanzee
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Cross Reactivity Sheep
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P04792
Host Species Mouse
Immunogen Partially purified hsp27 (earlier called 24K) protein from breast cancer MCF-7 cells.
Isotype IgG1, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3315
Reconstitution Protein A purified