Please login to your online account to display your discounted pricing

Mouse anti-HSP27, Clone: SPM252, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen HSP27
Clone SPM252
Concentration 1mg/mL
Immunogen Partially purified hsp27 (earlier called 24K) protein from breast cancer MCF-7 cells.
Gene Accession No. P04792
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23475101

 
novus biologicals canada
NBP23475101MG
0.1mg Each for $384.00

NBP23475102

 
novus biologicals canada
NBP23475102MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen HSP27
Clone SPM252
Concentration 1mg/mL
Immunogen Partially purified hsp27 (earlier called 24K) protein from breast cancer MCF-7 cells.
Gene Accession No. P04792
Gene ID (Entrez) 3315
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Chicken
Cross Reactivity Chimpanzee
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Sheep
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting