Please login to your online account to display your discounted pricing

Mouse anti-HSP27, Clone: SPM252, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23475102MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23475102

  • / Each
 

Description & Specifications

Specifications

Immunogen Partially purified hsp27 (earlier called 24K) protein from breast cancer MCF-7 cells.
Quantity 0.2mg
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Chimpanzee
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Sheep
Antigen HSP27
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3315
Test Specificity It recognizes a 24-27kDa estrogen-regulated protein, identified as heat shock protein 27 (hsp27).
Reconstitution Protein A purified
Clone SPM252
Concentration 1mg/mL
Gene Accession No. P04792