missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-HSP27, Clone: G3.1, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen HSP27
Clone G3.1
Concentration 1mg/mL
Immunogen Partially purified hsp27 (earlier called 24K) protein from breast cancer MCF-7 cells.
Gene Accession No. P04792
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23462101 Novus Biologicals Canada
NBP23462101MG
0.1mg Each for $384.00
NBP23462102 Novus Biologicals Canada
NBP23462102MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

HSP27
1mg/mL
P04792
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
G3.1
Partially purified hsp27 (earlier called 24K) protein from breast cancer MCF-7 cells.
3315
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot