Please login to your online account to display your discounted pricing

Mouse anti-HSP27, Clone: G3.1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23462102MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23462102

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Chicken
Cross Reactivity Chimpanzee
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Cross Reactivity Sheep
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Mouse
Conjugate Unlabeled
Immunogen Partially purified hsp27 (earlier called 24K) protein from breast cancer MCF-7 cells.
Regulatory Status RUO
Clone G3.1
Antigen HSP27
Gene Accession No. P04792
Primary or Secondary Primary
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Quantity 0.2mg
Isotype IgG1, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3315
Reconstitution Protein A purified
Concentration 1mg/mL
Purification Method Purified