Please login to your online account to display your discounted pricing

Mouse anti-HSP27, Clone: G3.1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23462101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23462101

  • / Each
 

Description & Specifications

Specifications

Antigen HSP27
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone G3.1
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P04792
Host Species Murine
Immunogen Partially purified hsp27 (earlier called 24K) protein from breast cancer MCF-7 cells.
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3315
Reconstitution Protein A purified
Target Species Chicken
Target Species Chimpanzee
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat
Target Species Sheep