Please login to your online account to display your discounted pricing

Mouse anti-HOP, Clone: DS14F5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP197503005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19750305

  • / Each
 

Description & Specifications

Specifications

Antigen HOP
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone DS14F5
Conjugate Unlabeled
Format Purified
Formulation Liquid. In PBS, pH 7.2 containing 50% glycerol. Protein G purified.
Host Species Murine
Immunogen Chicken HOP (p60).
Isotype IgG1
Purification Method Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 84525
Target Species Bovine
Target Species Canine
Target Species Chicken
Target Species Guinea Pig
Target Species Human
Target Species Mink
Target Species Murine
Target Species Porcine
Target Species Primate
Target Species Rabbit
Target Species Rat
Target Species Sheep
Target Species Xenopus