Please login to your online account to display your discounted pricing

Mouse anti-HO-1/HMOX1/HSP32, Clone: HO-1-2, PE, Novus Biologicals™

$505.00 - $505.00

Specifications

Antigen HO-1/HMOX1/HSP32
Clone HO-1-2
Immunogen Native rat HO-1.
Gene Accession No. P09601
Gene ID (Entrez) 3162
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP19746102

 
novus biologicals canada
NBP19746102MG
0.2mg Each for $505.00
Description & Specifications

Specifications

Antigen HO-1/HMOX1/HSP32
Clone HO-1-2
Immunogen Native rat HO-1.
Gene Accession No. P09601
Gene ID (Entrez) 3162
Formulation PBS containing 50% glycerol. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG2b
Conjugate PE
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Western Blot