Please login to your online account to display your discounted pricing

Mouse anti-HO-1/HMOX1/HSP32, Clone: HO-1-2, FITC, Novus Biologicals™

$505.00 - $505.00

Specifications

Antigen HO-1/HMOX1/HSP32
Clone HO-1-2
Immunogen Native rat HO-1.
Gene ID (Entrez) 3162
Formulation Liquid. In PBS. Protein G purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP19746802

 
novus biologicals canada
NBP19746802MG
0.2mg Each for $505.00
Description & Specifications

Specifications

Antigen HO-1/HMOX1/HSP32
Clone HO-1-2
Immunogen Native rat HO-1.
Gene ID (Entrez) 3162
Formulation Liquid. In PBS. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Canine
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Isotype IgG2b
Conjugate FITC
Format Purified
Purification Method Purified
Applications Flow Cytometry