Please login to your online account to display your discounted pricing

Mouse anti-HO-1/HMOX1/HSP32, Clone: HO-1-2, Biotin, Novus Biologicals™

$505.00 - $505.00

Specifications

Antigen HO-1/HMOX1/HSP32
Clone HO-1-2
Immunogen Native rat HO-1.
Gene ID (Entrez) 3162
Formulation Liquid. In PBS containing 50% glycerol. Protein G purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP19746402

 
novus biologicals canada
NBP19746402MG
0.2mg Each for $505.00
Description & Specifications

Specifications

Antigen HO-1/HMOX1/HSP32
Clone HO-1-2
Immunogen Native rat HO-1.
Gene ID (Entrez) 3162
Formulation Liquid. In PBS containing 50% glycerol. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Rat
Isotype IgG2b
Conjugate Biotin
Format Purified
Purification Method Purified
Applications Western Blotting