Please login to your online account to display your discounted pricing

Mouse anti-HLA G, Clone: 87G, Novus Biologicals™

$214.00 - $274.59

Specifications

Antigen HLA G
Clone 87G
Concentration 1.0mg/mL
Immunogen 721.221 cells transfected with full length HLA G cDNA with high surface HLA G expression. [UniProt No. P17693]
Gene Accession No. P17693
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14312303

 
novus biologicals canada
NBP1431230025MG
0.025mg Each for $214.00

NBP14312301

 
novus biologicals canada
NBP14312301MG
0.1mg Each for $274.59
Description & Specifications

Specifications

Antigen HLA G
Clone 87G
Concentration 1.0mg/mL
Immunogen 721.221 cells transfected with full length HLA G cDNA with high surface HLA G expression. [UniProt No. P17693]
Gene Accession No. P17693
Gene ID (Entrez) 3135
Formulation PBS. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Blocking
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Neutralization
Applications Western Blotting