Please login to your online account to display your discounted pricing

Mouse anti-HLA-DR, Clone: SPM289, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23475601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23475601

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P01903
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Format Purified
Immunogen Activated human peripheral blood mononuclear cells
Isotype IgG2b, κ
Purification Method Purified
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Gene ID (Entrez) 3122
Test Specificity This MAb reacts with the beta-chain of HLA-DR antigen, a member of MHC class II molecules. It does not cross react with HLA-DP and HLA-DQ. The L243 antibody recognizes a different epitope than the LN3 monoclonal antibody, and these antibodies do not cross-block binding to each other's respective epitopes. HLA-DR is a heterodimeric cell surface glycoprotein comprised of a 36kDa alpha (heavy) chain and a 28kDa beta (light) chain. It is expressed on B-cells, activated T-cells, monocytes/macrophages, dendritic cells and other non-professional APCs. In conjunction with the CD3/TCR complex and CD4 molecules, HLA-DR is critical for efficient peptide presentation to CD4+ T cells. It is an excellent histiocytic marker in paraffin sections producing intense staining. True histiocytic neoplasms are similarly positive. HLA-DR antigens also occur on a variety of epithelial cells and their corresponding neoplastic counterparts.
Reconstitution Protein A purified
Clone SPM289
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Antigen HLA-DR
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal