Please login to your online account to display your discounted pricing

Mouse anti-HLA-DR, Clone: LN3, PE-Cy7, Novus Biologicals™

$197.00 - $395.00

Specifications

Antigen HLA-DR
Clone LN3
Immunogen The immunogen for this antibody is HLA DR.
Gene ID (Entrez) 3122
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200284

 
novus biologicals canada
NBP200284100TESTS
100 Tests Each for $395.00

NBP20028425

 
novus biologicals canada
NBP20028425TESTS
25 Tests Each for $197.00
Description & Specifications

Specifications

Antigen HLA-DR
Clone LN3
Immunogen The immunogen for this antibody is HLA DR.
Gene ID (Entrez) 3122
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG2b, κ
Conjugate PE-Cy7
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry