Please login to your online account to display your discounted pricing

Mouse anti-HLA-DR, Clone: LN-3, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen HLA-DR
Clone LN-3
Concentration 0.2mg/mL
Immunogen Activated human peripheral blood mononuclear cells
Gene Accession No. P01903
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23446401

 
novus biologicals canada
NBP23446401ML
0.1mL Each for $139.00

NBP23446405

 
novus biologicals canada
NBP23446405ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen HLA-DR
Clone LN-3
Concentration 0.2mg/mL
Immunogen Activated human peripheral blood mononuclear cells
Gene Accession No. P01903
Gene ID (Entrez) 3122
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Primate
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting