Please login to your online account to display your discounted pricing

Mouse anti-HLA-DR, Clone: LN-3, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen HLA-DR
Clone LN-3
Concentration 1mg/mL
Immunogen Activated human peripheral blood mononuclear cells
Gene Accession No. P01903
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23467601

 
novus biologicals canada
NBP23467601MG
0.1mg Each for $384.00

NBP23467602

 
novus biologicals canada
NBP23467602MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen HLA-DR
Clone LN-3
Concentration 1mg/mL
Immunogen Activated human peripheral blood mononuclear cells
Gene Accession No. P01903
Gene ID (Entrez) 3122
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Primate
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting