Please login to your online account to display your discounted pricing

Mouse anti-HLA ABC, Clone: W6/32, PE-Cy5, Novus Biologicals™

$171.00 - $354.00

Specifications

Antigen HLA ABC
Clone W6/32
Immunogen Purified human tonsil lymphocyte membranes.
Gene ID (Entrez) 3105
Formulation Protein G purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200439

 
novus biologicals canada
NBP200439100TESTS
100 Tests Each for $354.00

NBP20043925

 
novus biologicals canada
NBP20043925TESTS
25 Tests Each for $171.00
Description & Specifications

Specifications

Antigen HLA ABC
Clone W6/32
Immunogen Purified human tonsil lymphocyte membranes.
Gene ID (Entrez) 3105
Formulation Protein G purified
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Baboon
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Primate
Isotype IgG2a
Conjugate PE-Cy5
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)