Please login to your online account to display your discounted pricing

Mouse anti-HLA ABC, Clone: W6/32, PE-Cy5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200439100TESTS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP200439

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation Protein G purified
Host Species Mouse
Antigen HLA ABC
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Immunogen Purified human tonsil lymphocyte membranes.
Quantity 100 Tests
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3105
Test Specificity NB100-64775 recognizes an antigenic determinant shared among products of the HLA A, B and C loci. Clone W6/32 recognizes a conformational epitope, reacting with HLA class I alpha3 and alpha2 domains (6). MHC class I glycoproteins are found on the surface of almost all nucleated somatic cells. This product is routinely tested in flow cytometry on human peripheral blood lymphocytes. Does not cross react with Amphibian, Canine, Chicken, Guinea Pig, Goat, Mouse or Rabbit.
Conjugate PE-Cy5
Cross Reactivity Baboon
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Primate
Isotype IgG2a
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Clone W6/32