Please login to your online account to display your discounted pricing

Mouse anti-HLA ABC, Clone: W6/32, FITC, Novus Biologicals™

$175.00 - $329.00

Specifications

Antigen HLA ABC
Clone W6/32
Immunogen Purified human tonsil lymphocyte membranes.
Gene ID (Entrez) 3105
Formulation Protein G purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200438

 
novus biologicals canada
NBP200438100TESTS
100 Tests Each for $329.00

NBP20043825

 
novus biologicals canada
NBP20043825TESTS
25 Tests Each for $175.00
Description & Specifications

Specifications

Antigen HLA ABC
Clone W6/32
Immunogen Purified human tonsil lymphocyte membranes.
Gene ID (Entrez) 3105
Formulation Protein G purified
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C in the dark.
Host Species Mouse
Cross Reactivity Baboon
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Primate
Isotype IgG2a
Conjugate FITC
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)