Please login to your online account to display your discounted pricing

Mouse anti-HIV Type 1, Clone: HIV1-24/661, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen HIV Type 1
Clone HIV1-24/661
Concentration 0.2mg/mL
Immunogen Recombinant HIV-1 Gag p24 protein
Gene Accession No. P04585
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23433505

 
novus biologicals canada
NBP23433505ML
0.5mL Each for $321.00

NBP23433501

 
novus biologicals canada
NBP23433501ML
0.1mL Each for $139.00
Description & Specifications

Specifications

Antigen HIV Type 1
Clone HIV1-24/661
Concentration 0.2mg/mL
Immunogen Recombinant HIV-1 Gag p24 protein
Gene Accession No. P04585
Gene ID (Entrez) 155348
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Virus
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence